missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Silica gel, for chromatography, 0.060-0.200 mm, 60 A

For column chromatography. | CAS: 7631-86-9 | O2Si | 60.08 g/mol

$1016.94 - $1016.94

Chemical Identifiers

CAS 7631-86-9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC240370050
SDS View Documents
Thermo Scientific Chemicals
240370050
5 kg
Each for $1,016.94
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7631-86-9

Specifications

Plastic Drum
Silica Gel
Column Chromatography
5 kg
Silica Gel
7631-86-9
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only