Learn More
Cytiva AcroPrep™ 96 Filter Plates, 1 mL - 3.0 m, glass fiber media/0.2 m Bio-Inert membrane (5/pkg)
Catalog No. NC1802620
Encompass
Optimized for a Variety of Applications Requiring Larger Per Well Volume ProcessingPlates are constructed of chemically resistant, biologically-inert polypropyleneRobotics-friendly design has single piece construction, designed in accordance with the ANSI/SBS X-2004 specificationsA serialized barcode label allows use in automated tracking systemsProprietary sealing technology individually seals each well, eliminating crosstalkAvailable in a variety of membrane configurationsExtended flow directors eliminate splattering and crosstalk of filtrate This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1802620 Supplier Cytiva Supplier No. 5053
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Optimized for a Variety of Applications Requiring Larger Per Well Volume ProcessingPlates are constructed of chemically resistant, biologically-inert polypropyleneRobotics-friendly design has single piece construction, designed in accordance with the ANSI/SBS X-2004 specificationsA serialized barcode label allows use in automated tracking systemsProprietary sealing technology individually seals each well, eliminating crosstalkAvailable in a variety of membrane configurationsExtended flow directors eliminate splattering and crosstalk of filtrate This product is not approved for use in medical, clinical, surgical or other patient applications.