missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein Dimethyl Acetal 97.0+%, TCI America™
$260.97 - $1286.19
Chemical Identifiers
| CAS | 6044-68-4 |
|---|---|
| Molecular Formula | C5H10O2 |
| Molecular Weight (g/mol) | 102.133 |
| MDL Number | MFCD00008632 |
| InChI Key | OBWGMYALGNDUNM-UHFFFAOYSA-N |
| Synonym | 3,3-Dimethoxy-1-propene |
| PubChem CID | 80145 |
| IUPAC Name | 3,3-dimethoxyprop-1-ene |
| SMILES | COC(C=C)OC |
Chemical Identifiers
| 6044-68-4 | |
| 102.133 | |
| OBWGMYALGNDUNM-UHFFFAOYSA-N | |
| 80145 | |
| COC(C=C)OC |
| C5H10O2 | |
| MFCD00008632 | |
| 3,3-Dimethoxy-1-propene | |
| 3,3-dimethoxyprop-1-ene |
Specifications
| 6044-68-4 | |
| 90°C | |
| MFCD00008632 | |
| 1993 | |
| OBWGMYALGNDUNM-UHFFFAOYSA-N | |
| 3,3-dimethoxyprop-1-ene | |
| 80145 | |
| ≥97.0% (GC) | |
| Acrolein Dimethyl Acetal |
| Colorless | |
| C5H10O2 | |
| 25 g | |
| 3,3-Dimethoxy-1-propene | |
| COC(C=C)OC | |
| 102.133 | |
| 102.13 | |
| Liquid |
Safety and Handling
RTECSNumber : UC8500000
TSCA : No