missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acrolein Diethyl Acetal 97.0+%, TCI America™
$200.21 - $902.90
Chemical Identifiers
| CAS | 3054-95-3 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.187 |
| MDL Number | MFCD00009239 |
| InChI Key | MCIPQLOKVXSHTD-UHFFFAOYSA-N |
| Synonym | acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal |
| PubChem CID | 62477 |
| IUPAC Name | 3,3-diethoxyprop-1-ene |
| SMILES | CCOC(C=C)OCC |
Chemical Identifiers
| 3054-95-3 | |
| 130.187 | |
| MCIPQLOKVXSHTD-UHFFFAOYSA-N | |
| 62477 | |
| CCOC(C=C)OCC |
| C7H14O2 | |
| MFCD00009239 | |
| acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal | |
| 3,3-diethoxyprop-1-ene |
Specifications
| 3054-95-3 | |
| 124°C | |
| MFCD00009239 | |
| 2374 | |
| MCIPQLOKVXSHTD-UHFFFAOYSA-N | |
| 3,3-diethoxyprop-1-ene | |
| 62477 | |
| ≥97.0% (GC) | |
| Acrolein Diethyl Acetal |
| Colorless | |
| C7H14O2 | |
| 25 g | |
| acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal | |
| CCOC(C=C)OCC | |
| 130.187 | |
| 130.19 | |
| Liquid |
Safety and Handling
RTECSNumber : AS1370000
TSCA : No