missing translation for 'onlineSavingsMsg'
Learn More

Acrolein Diethyl Acetal 97.0+%, TCI America™

Catalog No. A064025G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A064025G 25 g
A0640250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A064025G Supplier TCI America Supplier No. A064025G

Chemical Identifiers

CAS 3054-95-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.187
MDL Number MFCD00009239
InChI Key MCIPQLOKVXSHTD-UHFFFAOYSA-N
Synonym acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal
PubChem CID 62477
IUPAC Name 3,3-diethoxyprop-1-ene
SMILES CCOC(C=C)OCC

Specifications

Color Colorless
Boiling Point 124°C
Quantity 25 g
UN Number 2374
Formula Weight 130.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Acrolein Diethyl Acetal
RTECSNumber AS1370000
TSCA No