Learn More
Acrolein diethyl acetal, 94%
CAS: 3054-95-3 | C7H14O2 | 130.19 g/mol
$133.97 - $384.61
Chemical Identifiers
| CAS | 3054-95-3 |
|---|---|
| Molecular Formula | C7H14O2 |
| Molecular Weight (g/mol) | 130.19 |
| MDL Number | MFCD00009239 |
| InChI Key | MCIPQLOKVXSHTD-UHFFFAOYSA-N |
| Synonym | acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal |
| PubChem CID | 62477 |
| IUPAC Name | 3,3-diethoxyprop-1-ene |
| SMILES | CCOC(C=C)OCC |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC102770250
|
Thermo Scientific Chemicals
102770250 |
25 g | Glass bottle |
Each for $133.97
|
|
||||
|
AC102771000
|
Thermo Scientific Chemicals
102771000 |
100 g | Glass bottle |
Each for $384.61
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 3054-95-3 | |
| 130.19 | |
| MCIPQLOKVXSHTD-UHFFFAOYSA-N | |
| 62477 | |
| CCOC(C=C)OCC |
| C7H14O2 | |
| MFCD00009239 | |
| acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal | |
| 3,3-diethoxyprop-1-ene |
Specifications
| 3054-95-3 | |
| 100.0 | |
| 0.8500g/mL | |
| 15°C | |
| 92% min. (GC) | |
| C7H14O2 | |
| H2C=CHCH(OC2H5)2 | |
| 25 g | |
| 09,158 | |
| acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal | |
| MCIPQLOKVXSHTD-UHFFFAOYSA-N | |
| 3,3-diethoxyprop-1-ene | |
| 62477 | |
| 94% | |
| Acrolein diethyl acetal |
| 92.0 | |
| Colorless | |
| 120.0°C to 125.0°C | |
| Authentic | |
| Glass bottle | |
| 1.3970 to 1.4030 | |
| MFCD00009239 | |
| 01, 727 | |
| 0.85 | |
| Solubility in water: .. Other solubilities: miscible in ethanol,diethyl ether | |
| CCOC(C=C)OCC | |
| 130.19 | |
| 130.19 | |
| Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
GHS Signal Word: Danger
EINECSNumber : 221-276-
RUO – Research Use Only