missing translation for 'onlineSavingsMsg'
Learn More

Acrolein diethyl acetal, 94%

Catalog No. AC102771000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102771000 100 g Glass bottle
AC102770250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102771000 Supplier Thermo Scientific Chemicals Supplier No. 102771000

CAS: 3054-95-3 | C7H14O2 | 130.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3054-95-3
Molecular Formula C7H14O2
Molecular Weight (g/mol) 130.19
MDL Number MFCD00009239
InChI Key MCIPQLOKVXSHTD-UHFFFAOYSA-N
Synonym acrolein diethyl acetal, acrolein diethylacetal, acrolein acetal, 3,3-diethoxy-1-propene, 1-propene, 3,3-diethoxy, propenal diethyl acetal, 3,3-diethoxypropene, 1,1-diethoxy-2-propene, acrolein, diethyl acetal, acrylaldehyde diethyl acetal
PubChem CID 62477
IUPAC Name 3,3-diethoxyprop-1-ene
SMILES CCOC(C=C)OCC

Specifications

CAS Min % 92.0
CAS Max % 100.0
Color Colorless
Density 0.8500g/mL
Boiling Point 120.0°C to 125.0°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH(OC2H5)2
Refractive Index 1.3970 to 1.4030
Quantity 100 g
Beilstein 01, 727
Fieser 09,158
Specific Gravity 0.85
Solubility Information Solubility in water: .. Other solubilities: miscible in ethanol,diethyl ether
Formula Weight 130.19
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Acrolein diethyl acetal
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
EINECSNumber 221-276-

RUO – Research Use Only