missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acriflavine hydrochloride, MP Biomedicals™
$359.09 - $359.09
Chemical Identifiers
| CAS | 8063-24-9 |
|---|---|
| Molecular Formula | C27H27Cl3N6 |
| Molecular Weight (g/mol) | 541.91 |
| MDL Number | MFCD00069039 |
| InChI Key | MKLTXAHQKDVBLY-UHFFFAOYSA-N |
| Synonym | Euflavin, Trypaflavine |
| IUPAC Name | 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride |
| SMILES | Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19067580
|
MP Biomedicals Inc
0219067580 |
100 g |
N/A
|
|
|||||
Chemical Identifiers
| 8063-24-9 | |
| 541.91 | |
| MKLTXAHQKDVBLY-UHFFFAOYSA-N | |
| 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride |
| C27H27Cl3N6 | |
| MFCD00069039 | |
| Euflavin, Trypaflavine | |
| Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 |
Specifications
| 8063-24-9 | |
| C27H27Cl3N6 | |
| Euflavin, Trypaflavine | |
| Cl.Cl.[Cl-].NC1=CC2=NC3=CC(N)=CC=C3C=C2C=C1.C[N+]1=C2C=C(N)C=CC2=CC2=CC=C(N)C=C12 | |
| 541.91 | |
| Brown/Red | |
| Powder |
| 0.991 | |
| MFCD00069039 | |
| MKLTXAHQKDVBLY-UHFFFAOYSA-N | |
| 3,6-diamino-10-methylacridin-10-ium acridine-3,6-diamine dihydrochloride chloride | |
| 245.8 | |
| 100 g | |
| Acriflavine hydrochloride, Intercalating dye, antiseptic, analysis of cellular DNA and protein |
Safety and Handling
Recommended Storage : Store at room temperature.