missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acridine 98.0+%, TCI America™
Supplier: TCI America A02945G
Specifications
| Acridine | |
| 108°C | |
| 346°C | |
| MFCD00005025 | |
| 2713 | |
| DZBUGLKDJFMEHC-UHFFFAOYSA-N | |
| acridine | |
| 9215 | |
| 179.22 | |
| Crystalline Powder |
| 260-94-6 | |
| Yellow | |
| C13H9N | |
| 5 g | |
| 9-azaanthracene, 2,3-benzoquinoline, acrydine, 10-azaanthracene, akridin, dibenzo b,e pyridine, 2,3,5,6-dibenzopyridine, akridin czech, benzo b quinoline | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=N2 | |
| 179.222 | |
| CHEBI:36420 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 260-94-6 | |
| 179.222 | |
| DZBUGLKDJFMEHC-UHFFFAOYSA-N | |
| 9215 | |
| acridine |
| C13H9N | |
| MFCD00005025 | |
| 9-azaanthracene, 2,3-benzoquinoline, acrydine, 10-azaanthracene, akridin, dibenzo b,e pyridine, 2,3,5,6-dibenzopyridine, akridin czech, benzo b quinoline | |
| CHEBI:36420 | |
| C1=CC=C2C(=C1)C=C3C=CC=CC3=N2 |
Safety and Handling
RTECSNumber : AR7175000
TSCA : Yes