missing translation for 'onlineSavingsMsg'
Learn More

Acridine Orange

Catalog No. AAL1315906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1315906 5 g
AAL1315914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1315906 Supplier Thermo Scientific Chemicals Supplier No. L1315906
Only null left

CAS: 10127-02-3 | C17H20Cl3N3Zn | 438.10 g/mol

Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fluorescent stain for nucleic acids. An RNA polymerase inhibitorAcridine Orange acts as a lysosomal dye. It is used for cell-cycle studies. It plays an important role as a nucleic acid-selective fluorescent cationic dye, which is useful for cell cycle determination. Further, it serves as an inhibitor of RNA polymerase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10127-02-3
Molecular Formula C17H20Cl3N3Zn
Molecular Weight (g/mol) 438.10
MDL Number MFCD00081043,MFCD00081043,MFCD00081043
InChI Key VADJQOXWNSPOQA-UHFFFAOYSA-L
Synonym acridine orange c.i. 46005
PubChem CID 71774334
IUPAC Name zinc;hydron;3-N,3-N,6-N,6-N-tetramethylacridine-3,6-diamine;trichloride
SMILES [H+].[Cl-].[Cl-].[Cl-].[Zn++].CN(C)C1=CC2=NC3=CC(=CC=C3C=C2C=C1)N(C)C

Specifications

Quantity 5 g
Beilstein 3734978
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 438.1
Chemical Name or Material Acridine Orange
Hazard Category H302+H312+H332-H341
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
EINECSNumber 233-353-6
RTECSNumber AR7600000
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only