Learn More
Waters Limited ACQUITY UPLC Protein BEH SEC Guard Column, 200, 1.7 m, 4.6 mm X 30 mm, 10K - 500K, 1/pk
Supplier: Waters Limited 186005793
This guard column kit contains an advanced, ultra-low volume guard device (4.6 mm ID x 30 mm) containing BEH200, SEC, 1.7m particles and guard connection tubing (0.005 x 1.75) designed specifically for use in UPLC SEC applications up to 6000 psi (400 bar).