Learn More
Waters Limited ACQUITY UPLC BEH Phenyl Column, 130, 1.7 m, 3 mm X 150 mm, 1/pk
Catalog No. 50791183
Encompass
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemiscally stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.
Catalog No. 50791183 Supplier Waters Limited Supplier No. 186004674
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ACQUITY BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This sorbent, along with the XBridge BEH Phenyl, provide you with the most chemiscally stable phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape.