Learn More
Waters Limited ACQUITY UPLC / UV Qualification Standards Kit 25 mm Flow Cell
Catalog No. 50792806
Encompass
ACQUITY UPLC standards kit for system verification. Designed for use on UV detectors with 25mm Flow Cell. Kit contains standards appropriate for verification of system reproducibility, flow rate, gradient performance, detector linearity, detector accuracy, injector accuracy,injector carryover.
Catalog No. 50792806 Supplier Waters Limited Supplier No. 700002846
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
ACQUITY UPLC standards kit for system verification. Designed for use on UV detectors with 25mm Flow Cell. Kit contains standards appropriate for verification of system reproducibility, flow rate, gradient performance, detector linearity, detector accuracy, injector accuracy,injector carryover.