Learn More
Waters Limited ACQUITY UPLC Oligonucleotide BEH C18 Method Validation Kit, 130, 1.7 m, 2.1 mm X 100 mm, 3/pk
Catalog No. 50466121
Encompass
The reproducibility of a chromatographic column has a critical impact on the long-term reliability and robustness of an analytical method, and its reproducibility lies completely outside the user's control. With exceptional batch-to-batch and column-to-column reproducibility, Waters well-established particle and column manufacturing process control provides confidence in the long-term reliability of your analytical method. ACQUITY OST C18, 135A, 1.7um Method Validation Kit includes three batches of chromatographic media [derived from different base particles] to judge the quality, reliability, and consistency of your chromatographic method. Each batch of selected ACQUITY OST C18, 135A, 1.7um chemistry is specifically QC tested with a mixture of synthetic oligonucleotides to help ensure consistent performance.
Catalog No. 50466121 Supplier Waters Limited Supplier No. 186004898
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The reproducibility of a chromatographic column has a critical impact on the long-term reliability and robustness of an analytical method, and its reproducibility lies completely outside the user's control. With exceptional batch-to-batch and column-to-column reproducibility, Waters well-established particle and column manufacturing process control provides confidence in the long-term reliability of your analytical method. ACQUITY OST C18, 135A, 1.7um Method Validation Kit includes three batches of chromatographic media [derived from different base particles] to judge the quality, reliability, and consistency of your chromatographic method. Each batch of selected ACQUITY OST C18, 135A, 1.7um chemistry is specifically QC tested with a mixture of synthetic oligonucleotides to help ensure consistent performance.