Learn More
Waters Limited ACQUITY UPLC CSH Fluoro-Phenyl VanGuard Pre-column, 130, 1.7 m, 2.1 mm X 5 mm, 3/pk
Catalog No. 50791377
Encompass
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds. The VanGuard pre-column connects directly to the inlet of the ACQUITY UPLC column which minimizes extra columns volumes and reduces leaking by eliminating additional connections.
Catalog No. 50791377 Supplier Waters Limited Supplier No. 186005358
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to maximize selectivity differences, the ACQUITY CSH Fluoro-Phenyl column provides maximum method development freedom, especially when using low pH mobile phases. The non-encapped sorbent provides you with enhanced retention of acidic compounds. The VanGuard pre-column connects directly to the inlet of the ACQUITY UPLC column which minimizes extra columns volumes and reduces leaking by eliminating additional connections.