missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acidity, MilliporeSigma™ Supelco™
Description
Featured Industries: Environmental and Food and Beverages
Specifications
Specifications
| CAS | 7647-01-0 |
| Type | Certified Reference Material |
| Quantity | 20 mL |
| Grade | Certified Reference Material |