missing translation for 'onlineSavingsMsg'
Learn More

Acid Yellow 3, TCI America™

Catalog No. Q002325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q002325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q002325G Supplier TCI America Supplier No. Q002325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

  • Chemical Name: Acid Yellow 3
  • Physical Form: Crystal/Powder
  • Color: Yellow to reddish yellow
  • Molecular Formula: C18H9NNa2O8S2
  • Molecular Weight: 477.37
  • Solubility: Soluble in water; slightly soluble in ethanol, glycerol, and glycol
  • Maximum Absorption Wavelength: 412 nm
  • Lambda max.: 410.0 to 414.0 nm (H2O)
  • Absorbance (E1%1cm) min.: 600
  • Acute Toxicity (orl-rat LD50): 2 g/kg
  • Storage Temperature: Room Temperature
  • Color Index Number: 47005

Chemical Identifiers

CAS 8004-92-0
Molecular Formula C18H9NNa2O8S2
Molecular Weight (g/mol) 477.37
MDL Number MFCD00080727
InChI Key FZUOVNMHEAPVBW-UHFFFAOYSA-L
Synonym Quinaldine Yellow, Quinoline Yellow S
PubChem CID 129893443
IUPAC Name disodium 2-(1,3-dioxo-2,3-dihydro-1H-inden-2-yl)quinoline-6,8-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC(=C2N=C(C=CC2=C1)C1C(=O)C2=CC=CC=C2C1=O)S([O-])(=O)=O

Specifications

Color Yellow
Quantity 25 g
Physical Form Crystal/Powder
Grade Research
Chemical Name or Material Acid Yellow 3
EINECSNumber (5)-1393
RTECSNumber GC5796000
TSCA Yes