missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Acid-Resistant Biopsy Bags, Medium
Description
- Allow unrestricted fluid movement around tissue
- Reduce reagent carryover and pressure artifact often found with sponges
- Remove specimen rapidly with convenient peel-open bag
Dimensions:
- Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
- Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
- Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)
Specifications
Specifications
| Product Type | Biopsy Bag |
| Material | Nylon, Acid-resistant |
| Color | White |
| Length (Metric) | 31 mm |
| Width (Metric) | 43 mm |
| Length (English) | 1.2 in. |
| Width (English) | 1.7 in. |
| Size | Small |