missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Acid-Resistant Biopsy Bags, Medium

Catalog No. 2861316
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
2861316 Large
2861310 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 2861316 Supplier Epredia™ Supplier No. 6774052

Reduce risk of small specimen loss with Epredia™ Acid-Resistant Biopsy Bags.

  • Allow unrestricted fluid movement around tissue
  • Reduce reagent carryover and pressure artifact often found with sponges
  • Remove specimen rapidly with convenient peel-open bag

Dimensions:

  • Small Biopsy Bags measure 1.2 × 1.7″ (31 × 43mm)
  • Medium Biopsy Bags measure 1.8 × 2.6″ (45 × 65mm)
  • Large Biopsy Bags measure 3 × 3.5″ (75 × 85mm)

Specifications

Product Type Biopsy Bag
Material Nylon, Acid-resistant
Color White
Length (Metric) 75 mm
Width (Metric) 85 mm
Length (English) 3 in.
Width (English) 3.5 in.
Size Large