missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 94, TCI America™

Catalog No. R004125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R004125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R004125G Supplier TCI America Supplier No. R004125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 632-69-9
Molecular Formula C20H4Cl4I4Na2O5
Molecular Weight (g/mol) 1019.644
MDL Number MFCD00151169
InChI Key DPBQSRZQYAYDGY-UHFFFAOYSA-N
Synonym Rose Bengal
PubChem CID 87244310
IUPAC Name sodium;4,5,6,7-tetrachloro-3',6'-dihydroxy-2',4',5',7'-tetraiodospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=C2C(=C(C(=C1I)O)I)OC3=C(C(=C(C=C3C24C5=C(C(=C(C(=C5Cl)Cl)Cl)Cl)C(=O)O4)I)O)I.[Na].[Na]

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 1017.63
Chemical Name or Material Acid Red 94
EINECSNumber (4)-0650
RTECSNumber LM5920000
TSCA Yes