missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 88, TCI America™

Catalog No. F008725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F008725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F008725G Supplier TCI America Supplier No. F008725G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1658-56-6
Molecular Formula C20H13N2NaO4S
Molecular Weight (g/mol) 400.384
MDL Number MFCD00004070
InChI Key YCRBPWIFISBYGJ-UHFFFAOYSA-M
Synonym Echt Rot, Fast Red A, Roccelline
PubChem CID 23722700
IUPAC Name sodium;4-[2-(2-oxonaphthalen-1-ylidene)hydrazinyl]naphthalene-1-sulfonate
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C4=CC=CC=C43)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 280°C
Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 400.38
Chemical Name or Material Acid Red 88
EINECSNumber (5)-1512
RTECSNumber QK2420000
TSCA Yes