missing translation for 'onlineSavingsMsg'
Learn More

Acid Red 52 98.0+%, TCI America™

Catalog No. A060025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A060025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A060025G Supplier TCI America Supplier No. A060025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 3520-42-1
Molecular Formula C29H33N2NaO7S2
Molecular Weight (g/mol) 608.70
MDL Number MFCD00010180
InChI Key XJENLUNLXRJLEZ-UHFFFAOYSA-M
Synonym Acid Red, Sulforhodamine B, Xylene Red
PubChem CID 131852807
IUPAC Name sodium 3,6-bis(diethylamino)-9-(2,4-disulfonatophenyl)-2,7-dimethyl-10λ⁴-xanthen-10-ylium
SMILES [Na+].CCN(CC)C1=CC2=[O+]C3=CC(N(CC)CC)=C(C)C=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S([O-])(=O)=O)=C2C=C1C

Specifications

Quantity 25 g
Physical Form Crystal/Powder
Formula Weight 580.65
Percent Purity ≥98.0% (HPLC)
Chemical Name or Material Acid Red 52
EINECSNumber (5)-1504
RTECSNumber BP6750000
TSCA Yes