missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Red 26, TCI America™
Specifications
Specifications
| Color | Red |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 480.42 |
| Chemical Name or Material | Acid Red 26 |
Safety and Handling
| EINECSNumber | (5)-1496 |
| RTECSNumber | QJ6825000 |
| TSCA | Yes |