missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Red 18 85.0+%, TCI America™
Specifications
Specifications
| Color | Red |
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 604.46 |
| Percent Purity | ≥85.0% (E) |
| Chemical Name or Material | Acid Red 18 |
Safety and Handling
| EINECSNumber | (5)-1495 |
| RTECSNumber | QJ6530000 |
| TSCA | Yes |