missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Red 112 85.0+%, TCI America™
Supplier: TCI America A160025G
Specifications
| Acid Red 112 | |
| C22H12N4Na4O13S4 | |
| Ponceau S | |
| C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+] | |
| 760.552 | |
| 760.55 | |
| 25 g |
| 6226-79-5 | |
| MFCD00003892 | |
| VSXKEUCERCWGKF-STNZDNLRSA-J | |
| tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate | |
| 11320219 | |
| ≥85.0% (N) | |
| Crystal/Powder |
Chemical Identifiers
| 6226-79-5 | |
| 760.552 | |
| VSXKEUCERCWGKF-STNZDNLRSA-J | |
| 11320219 | |
| C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+] |
| C22H12N4Na4O13S4 | |
| MFCD00003892 | |
| Ponceau S | |
| tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate |
Safety and Handling
RTECSNumber : QJ6600000
TSCA : Yes