missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Red 112 85.0+%, TCI America™
Specifications
Specifications
| Formula Weight | 760.55 |
| Percent Purity | ≥85.0% (N) |
| Quantity | 1 g |
| Physical Form | Crystal/Powder |
| Chemical Name or Material | Acid Red 112 [for Biochemical Research] |
Safety and Handling
| RTECSNumber | QJ6600000 |
| TSCA | Yes |