missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LaMotte™ Acid Rain Study Outfit
Description
Includes
- Durable, tapered rain gauge
- Easy-to-use pH test kit
- Comprehensive study guide
- Activity sheets
- 50 tests
- Collect and measure precipitation and determine the pH levels of precipitation, ponds, lakes and streams with this complete outfit
- A valuable unit for an environmental science program
- Reagent refill available
Order Info
Grades 5+.
Specifications
Specifications
| Includes | Durable, tapered rain gauge, easy-to-use pH test kit, comprehensive study guide, activity sheets |
| Class Size | 1 Class |
| Product Type | Acid Rain Study Outfit |