missing translation for 'onlineSavingsMsg'
Learn More

LaMotte™ Acid Rain Study Outfit

Catalog No. S85000R
Change view
Click to view available options
Product Type:
Acid Rain Study Outfit
Acid Rain Study Outfit Refill
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S85000R Acid Rain Study Outfit Refill
S85000 Acid Rain Study Outfit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S85000R Supplier LaMotte™ Supplier No. 2218GACID

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Durable, tapered rain gauge
  • Easy-to-use pH test kit
  • Comprehensive study guide
  • Activity sheets
  • 50 tests

Tools for testing the acidity of rain.

  • Collect and measure precipitation and determine the pH levels of precipitation, ponds, lakes and streams with this complete outfit
  • A valuable unit for an environmental science program
  • Reagent refill available

Order Info

Grades 5+.

Specifications

Includes Durable, tapered rain gauge, easy-to-use pH test kit, comprehensive study guide, activity sheets
Class Size 1 Class
Product Type Acid Rain Study Outfit Refill