missing translation for 'onlineSavingsMsg'
Learn More

Acid Orange 7 97.0+%, TCI America™

Catalog No. p-7130167
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G015825G 25 g
G0158100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G015825G Supplier TCI America Supplier No. G015825G

Chemical Identifiers

CAS 633-96-5
Molecular Formula C16H11N2NaO4S
Molecular Weight (g/mol) 350.324
MDL Number MFCD00011657
InChI Key IGQFKZCLTLAWLO-UHFFFAOYSA-M
Synonym c.i. acid orange 7, orange ii, acid orange 7, orange, c.i. acid orange 7, monosodium salt, dsstox_cid_18881, dsstox_rid_79415, dsstox_gsid_38881, sodium 4-e-2-2-hydroxynaphthalen-1-yl diazen-1-yl benzenesulfonate, benzenesulfonic acid, 4-2-2-hydroxy-1-naphthalenyl diazenyl-, sodium salt 1:1
PubChem CID 44135675
IUPAC Name sodium;4-[2-(2-oxonaphthalen-1-ylidene)hydrazinyl]benzenesulfonate
SMILES C1=CC=C2C(=C1)C=CC(=O)C2=NNC3=CC=C(C=C3)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point 164°C
Quantity 25 g
Formula Weight 350.32
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Acid Orange 7
EINECSNumber (5)-1455
RTECSNumber DB7084000
TSCA Yes