missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Green 50, TCI America™
Specifications
Specifications
| Quantity | 25 g |
| Physical Form | Crystal/Powder |
| Formula Weight | 576.61 |
| Grade | Research |
| Chemical Name or Material | Acid Green 50 |
Safety and Handling
| EINECSNumber | (5)-1751 |
| RTECSNumber | BQ1150000 |
| TSCA | Yes |