missing translation for 'onlineSavingsMsg'
Learn More

Acid Green 1 80.0+%, TCI America™

Catalog No. N030625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N030625G 25 g
N0306500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N030625G Supplier TCI America Supplier No. N030625G

Chemical Identifiers

CAS 19381-50-1
Molecular Formula C30H18FeN3Na3O15S3
Molecular Weight (g/mol) 881.474
MDL Number MFCD00003886
InChI Key VLBLPLXOYXEXJK-UHFFFAOYSA-K
Synonym Naphthol Green B
PubChem CID 14598749
IUPAC Name trisodium;6-hydroxy-5-nitrosonaphthalene-2-sulfonate;iron
SMILES C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].C1=CC2=C(C=CC(=C2N=O)O)C=C1S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Fe]

Specifications

Formula Weight 878.45
Percent Purity ≥80.0% (E)
Color Black/Green
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Acid Green 1
EINECSNumber (5)-4373
RTECSNumber LJ8930000
TSCA Yes