missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Acid Fuchsin, pure, high purity biological stain

Catalog No. AC227900250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC227900250 25 g Glass bottle
AC227901000 100 g Glass bottle
AC227905000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC227900250 Supplier Thermo Scientific Chemicals Supplier No. 227900250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 3244-88-0 | C20H17N3Na2O9S3 | 587.544 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3244-88-0
Molecular Formula C20H17N3Na2O9S3
Molecular Weight (g/mol) 587.544
MDL Number MFCD00013286
InChI Key VCEFFPMSOGDMJX-UHFFFAOYSA-N
Synonym Acid Violet 19, C.I. 42685, Fuchsin Acid, Rubin S
PubChem CID 131852436
IUPAC Name 3-[bis(4-amino-3-sulfophenyl)methylidene]-6-imino-5-methylcyclohexa-1,4-diene-1-sulfonic acid;sodium
SMILES CC1=CC(=C(C2=CC(=C(C=C2)N)S(=O)(=O)O)C3=CC(=C(C=C3)N)S(=O)(=O)O)C=C(C1=N)S(=O)(=O)O.[Na].[Na]

Specifications

Absorbance 700 (at lambda min.)
Absorption Ratio 1 to 1.4
Solubility Information Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol
Infrared Spectrum Authentic
Lambda Max 544 to 550nm (in pH 3 acidified water)
Formula Weight 585.52
Grade Pure
Packaging Glass bottle
Color Green
Quantity 25 g
Physical Form Powder or Crystals
Chemical Name or Material Acid Fuchsin
Show More Show Less
EINECSNumber 221-816-5
RTECSNumber DD4737000
TSCA TSCA

RUO – Research Use Only