missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Acid Fuchsin, pure, certified
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Dye Content | 55% min. |
| Packaging | Glass bottle |
| Merck Index | 15, 99 |
| Color | Green |
| Solubility Information | Solubility in water: 143g/L. Other solubilities: slightly soluble in alcohol |
| Quantity | 25 g |
| Physical Form | Glittering Powder |
| Formula Weight | 585.52 |
| Grade | Pure |
| Chemical Name or Material | Acid Fuchsin, Pure |
RUO – Research Use Only