Learn More
StatLab™ Acid Fast Bacteria Tissue Control Slides
AFB Acid Fast Bacteria control slides consist of formalin fixed, paraffin embedded tissue sections which contain known distinctive tissue elements.
Supplier:  StatLab™ CSAFB25
Catalog No. STLCSAFB25
Only null left
Description
Description
  • Use these slides with confidence to evaluate stain quality and performance in histology laboratories
  • Each package includes specific staining protocol with stained slide included
  • Tissue: Known positive
  • Special Stain or Antibody: AFB Acid Fast Bacillus
Specifications
Specifications
Control Slide
25 Slides/Pk.
Product Suggestions
Product Suggestions