missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Acid Fast Bacillus Stain Kits
Produce optimal staining while reducing precipitate with the Epredia™ Acid Fast Bacillus.
Supplier: Epredia™ 87016
Description
Acid-fast bacilli are stained bright red against a pale blue or pale green background.Green Secondary Stain Kit includes:
- Fast Green Stain Solution, 250mL
- Carbol Fuchsin Solution, 125mL
- Differentiating Solution, 500mL
Blue Secondary Stain Kit includes:
- Loeffler Methylene Blue Stain Solution
- Carbol Fuchsin Solution, 125mL
- Differentiating Solution, 500mL
Specifications
| Acid Fast Bacillus, Blue Secondary Stain Kit |