missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Dichromate Cleaning Solution, Reagents
Specifications
Specifications
| Color-coded | No |
| Form | Liquid |
| pH | <1 |
| Sterility | Non-sterile |
| Autoclavable | Not Autoclavable |
| Boiling Point | 300°C |
| Color | Brown |
| Format | Liquid |
| Quantity | 1 L |
| Solubility | Completely miscible |
| Show More |