missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acid Blue 3 Sodium Salt, TCI America™
Supplier: TCI America A12425G
Specifications
| Acid Blue 3 Sodium Salt | |
| C27H31N2NaO7S2 | |
| PMLFOMWMYRKZRF-UHFFFAOYSA-M | |
| sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonate | |
| 9916325 | |
| 5 g |
| 20262-76-4 | |
| MFCD00012119 | |
| CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+] | |
| 582.662 | |
| 582.66 | |
| Crystal/Powder |
Chemical Identifiers
| 20262-76-4 | |
| 582.662 | |
| PMLFOMWMYRKZRF-UHFFFAOYSA-M | |
| sodium;4-[[4-(diethylamino)phenyl]-(4-diethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-6-hydroxybenzene-1,3-disulfonate |
| C27H31N2NaO7S2 | |
| MFCD00012119 | |
| 9916325 | |
| CCN(CC)C1=CC=C(C=C1)C(=C2C=CC(=[N+](CC)CC)C=C2)C3=CC(=C(C=C3S(=O)(=O)[O-])S(=O)(=O)[O-])O.[Na+] |
Safety and Handling
TSCA : No