missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Acid/Base Spill Kit

Catalog No. 19140941
Change view
Click to view available options
Capacity (English):
2.32 gal.
2.97 gal.
For Use With (Application):
Clean Acid and Base Spills
Clean Acid Spills
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) For Use With (Application)
19140941 2.97 gal. Clean Acid and Base Spills
19140935 2.32 gal. Clean Acid Spills
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19140941 Supplier Fisherbrand™ Supplier No. FSCSKCOMBO

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

2 lb. acid neutralizer shaker; 6 plastic locking bags with white write-on area; goggles' gloves; boot covers; apron; dust pan; hand broom; 2 lb. base neutralizer shaker; 3 lb. bag granular; spatula; plain label

An effective spill response solution in an easy-to-use kit

  • Chemical neutralizing polymers provide safe spill control with a color change indicator that reflects neutralization
  • Provides safe spill control for most acids
  • Improve worker productivity and efficiency by placing spill kits around your facility close to areas prone to spills

Compliance

OSHA 29 CFR 1910.22(a)(2); EPA 40 CFR 112.7(c)(1)(vii); OSHA 29 CFR 1910,120(j)(1)(vii).

Specifications

Capacity (English) Sorbing 2.97 gal.
Color White
Capacity (English) 2.97 gal.
Capacity (Metric) 38 L
Material Polyethylene
Type Universal Combo Spill Kit
For Use With (Application) Clean Acid and Base Spills
Includes 2 lb. acid neutralizer shaker, 6 plastic locking bags with white write-on area, goggles' gloves, boot covers, apron, dust pan, hand broom, 2 lb. base neutralizer shaker, 3 lb. bag granular, spatula, plain label
Height (English) 18 in.
Height (Metric) 45.7 cm