missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylthiocholine Iodide 98.0+%, TCI America™
$59.65 - $812.73
Chemical Identifiers
| CAS | 1866-15-5 |
|---|---|
| Molecular Formula | C7H16INOS |
| Molecular Weight (g/mol) | 289.175 |
| MDL Number | MFCD00011819 |
| InChI Key | NTBLZMAMTZXLBP-UHFFFAOYSA-M |
| Synonym | acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide |
| PubChem CID | 74629 |
| IUPAC Name | 2-acetylsulfanylethyl(trimethyl)azanium;iodide |
| SMILES | CC(=O)SCC[N+](C)(C)C.[I-] |
Chemical Identifiers
| 1866-15-5 | |
| 289.175 | |
| NTBLZMAMTZXLBP-UHFFFAOYSA-M | |
| 74629 | |
| CC(=O)SCC[N+](C)(C)C.[I-] |
| C7H16INOS | |
| MFCD00011819 | |
| acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide | |
| 2-acetylsulfanylethyl(trimethyl)azanium;iodide |
Specifications
| 1866-15-5 | |
| White | |
| MFCD00011819 | |
| 2811 | |
| NTBLZMAMTZXLBP-UHFFFAOYSA-M | |
| 2-acetylsulfanylethyl(trimethyl)azanium;iodide | |
| 74629 | |
| ≥98.0% (T) | |
| Acetylthiocholine Iodide |
| 206°C | |
| C7H16INOS | |
| 1 g | |
| acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide | |
| CC(=O)SCC[N+](C)(C)C.[I-] | |
| 289.175 | |
| 289.18 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : FZ9865000
TSCA : Yes