missing translation for 'onlineSavingsMsg'
Learn More

Acetylthiocholine Iodide 98.0+%, TCI America™

Catalog No. A01161G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A01161G 1 g
A01165G 5 g
A011625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A01161G Supplier TCI America Supplier No. A01161G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1866-15-5
Molecular Formula C7H16INOS
Molecular Weight (g/mol) 289.175
MDL Number MFCD00011819
InChI Key NTBLZMAMTZXLBP-UHFFFAOYSA-M
Synonym acetylthiocholine iodide, s-acetylthiocholine iodide, acetylthiocholine diiodide, 2-acetylthioethyltrimethylammonium iodide, 2-acetylthio-n,n,n-trimethylethanaminium iodide, ethanaminium, 2-acetylthio-n,n,n-trimethyl-, iodide, acetylthiocholinjodid, choline, s-acetylthio-, iodide, 2-acetylsulfanyl-n,n,n-trimethylethanaminium iodide, acetyl thiobilineurine iodide
PubChem CID 74629
IUPAC Name 2-acetylsulfanylethyl(trimethyl)azanium;iodide
SMILES CC(=O)SCC[N+](C)(C)C.[I-]

Specifications

Melting Point 206°C
Color White
Quantity 1 g
UN Number 2811
Formula Weight 289.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetylthiocholine Iodide
RTECSNumber FZ9865000
TSCA Yes