missing translation for 'onlineSavingsMsg'
Learn More

Acetylsalicylsalicylic acid, 97%

CAS: 530-75-6 | C16H12O6 | 300.26 g/mol

Supplier:  Thermo Scientific Chemicals 269350010

 View more versions of this product

Catalog No. AC269350010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetylsalicylsalicylic acid
530-75-6
100.0
96% min. (As is) Calculated (HPLC)
C16H12O6
1 g
14,1
Solubility in water: .. Other solubilities: soluble in alkaline solutions,soluble in alcohol,practically insoluble in benzene
CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=CC=C2C(=O)O
300.26
300.26
97%
96.0
145°C to 160°C
Glass bottle
MFCD00143537
10,IV,165
2-2-acetoxybenzoyl oxy benzoic acid, acetylsalicylsalicylic acid, acesalum, acetylsalicylsalicyic acid, diplosal acetate, unii-vbe72mcp5l, 2-carboxyphenyl 2-acetyloxy benzoate, diplosalsalate, ccris 1740, benzoic acid, 2-acetyloxy-, 2-carboxyphenyl ester
DDSFKIFGAPZBSR-UHFFFAOYSA-N
2-(2-acetyloxybenzoyl)oxybenzoic acid
10745
97%

Chemical Identifiers

530-75-6
300.26
DDSFKIFGAPZBSR-UHFFFAOYSA-N
10745
CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=CC=C2C(=O)O
C16H12O6
MFCD00143537
2-2-acetoxybenzoyl oxy benzoic acid, acetylsalicylsalicylic acid, acesalum, acetylsalicylsalicyic acid, diplosal acetate, unii-vbe72mcp5l, 2-carboxyphenyl 2-acetyloxy benzoate, diplosalsalate, ccris 1740, benzoic acid, 2-acetyloxy-, 2-carboxyphenyl ester
2-(2-acetyloxybenzoyl)oxybenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 208-493-6

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only