missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylenedicarboxylic Acid 90.0+%, TCI America™
$45.57 - $182.26
Chemical Identifiers
| CAS | 142-45-0 |
|---|---|
| Molecular Formula | C4H2O4 |
| Molecular Weight (g/mol) | 114.06 |
| MDL Number | MFCD00004362 |
| InChI Key | YTIVTFGABIZHHX-UHFFFAOYSA-N |
| Synonym | acetylenedicarboxylic acid, 2-butynedioic acid, butynedioic acid, acetylenedicarboxylate, unii-2d2oj4ko44, acetylenedicarboxylic acid 8ci, acetylendicarboxylate, hoocc=ccooh, acetylenedicarboxylicacid, pubchem13735 |
| PubChem CID | 371 |
| ChEBI | CHEBI:30781 |
| IUPAC Name | but-2-ynedioic acid |
| SMILES | OC(=O)C#CC(O)=O |
Chemical Identifiers
| 142-45-0 | |
| 114.06 | |
| YTIVTFGABIZHHX-UHFFFAOYSA-N | |
| 371 | |
| but-2-ynedioic acid |
| C4H2O4 | |
| MFCD00004362 | |
| acetylenedicarboxylic acid, 2-butynedioic acid, butynedioic acid, acetylenedicarboxylate, unii-2d2oj4ko44, acetylenedicarboxylic acid 8ci, acetylendicarboxylate, hoocc=ccooh, acetylenedicarboxylicacid, pubchem13735 | |
| CHEBI:30781 | |
| OC(=O)C#CC(O)=O |
Specifications
| 142-45-0 | |
| White-Yellow | |
| MFCD00004362 | |
| 2923 | |
| YTIVTFGABIZHHX-UHFFFAOYSA-N | |
| but-2-ynedioic acid | |
| 371 | |
| 114.06 | |
| Crystalline Powder |
| 178°C | |
| C4H2O4 | |
| 5 g | |
| acetylenedicarboxylic acid, 2-butynedioic acid, butynedioic acid, acetylenedicarboxylate, unii-2d2oj4ko44, acetylenedicarboxylic acid 8ci, acetylendicarboxylate, hoocc=ccooh, acetylenedicarboxylicacid, pubchem13735 | |
| OC(=O)C#CC(O)=O | |
| 114.06 | |
| CHEBI:30781 | |
| ≥90.0% (HPLC) | |
| Acetylenedicarboxylic Acid |
Safety and Handling
EINECSNumber : (2)-3393
TSCA : Yes
Recommended Storage : Refrigerator