missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylene-PEG4-maleimide, Thermo Scientific™
Chemical Identifiers
| CAS | 1609651-90-2 |
|---|---|
| Molecular Formula | C18H26N2O7 |
| Molecular Weight (g/mol) | 382.41 |
| MDL Number | MFCD22380740 |
| InChI Key | GHEBECZRHJXPTL-UHFFFAOYSA-N |
| Synonym | acetylene-peg4-maleimide, propargyl-peg4-maleimide, alkyne-peg4-maleimide hplc |
| PubChem CID | 91757777 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ64859MB
|
Thermo Scientific Chemicals
J64859MB |
25mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsHeterobifunctional cross linker enabling the connection of an azide containing compound or biomolecule to one with a thiol. The maleimide group will react preferentially with thiols to form a covalent adduct and the terminal alkyne will react with azides via copper-catalyzed 1,3-dipolar cycloaddition click chemistry. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification cysteine containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character. A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
| 1609651-90-2 | |
| 382.41 | |
| GHEBECZRHJXPTL-UHFFFAOYSA-N | |
| 91757777 |
| C18H26N2O7 | |
| MFCD22380740 | |
| acetylene-peg4-maleimide, propargyl-peg4-maleimide, alkyne-peg4-maleimide hplc |
Specifications
| 1609651-90-2 | |
| MFCD22380740 | |
| acetylene-peg4-maleimide, propargyl-peg4-maleimide, alkyne-peg4-maleimide hplc | |
| GHEBECZRHJXPTL-UHFFFAOYSA-N | |
| 91757777 | |
| Acetylene-PEG4-maleimide |
| C18H26N2O7 | |
| 25mg | |
| Soluble in water. | |
| 382.41 | |
| 382.41 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO â Research Use Only