missing translation for 'onlineSavingsMsg'
Learn More

Acetylcyclobutane 98.0+%, TCI America™

Catalog No. C10341ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10341ML 1 mL
C10345ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10341ML Supplier TCI America Supplier No. C10341ML
Only null left

Chemical Identifiers

CAS 3019-25-8
Molecular Formula C6H10O
Molecular Weight (g/mol) 98.145
MDL Number MFCD00037148
InChI Key JPJOOTWNILDNAW-UHFFFAOYSA-N
Synonym cyclobutyl methyl ketone, 1-cyclobutylethan-1-one, acetylcyclobutane, ethanone, 1-cyclobutyl, ketone, cyclobutyl methyl, methyl cyclobutyl ketone, cyclobutylmethylketone, acetylcyclobutan, cyclobutylethanone, cyclobutane, acetyl
PubChem CID 76398
IUPAC Name 1-cyclobutylethanone
SMILES CC(=O)C1CCC1

Specifications

Color Colorless
Boiling Point 137°C
Quantity 1 mL
UN Number 1224
Formula Weight 98.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Acetylcyclobutane