missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine Iodide 98.0+%, TCI America™

Catalog No. A008525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A008525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A008525G Supplier TCI America Supplier No. A008525G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2260-50-6
Molecular Formula C7H16INO2
Molecular Weight (g/mol) 273.11
MDL Number MFCD00011815
InChI Key SMBBQHHYSLHDHF-UHFFFAOYSA-M
Synonym acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
PubChem CID 75271
IUPAC Name [2-(acetyloxy)ethyl]trimethylazanium iodide
SMILES [I-].CC(=O)OCC[N+](C)(C)C

Specifications

Melting Point 162°C
Color White
Quantity 25 g
Formula Weight 273.11
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetylcholine Iodide
EINECSNumber (9)-1504
RTECSNumber KH3300000
TSCA Yes