missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine iodide, 99%

Catalog No. AC400180100
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC400180100 10 g
AC400180500 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400180100 Supplier Thermo Scientific Chemicals Supplier No. 400180100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2260-50-6 | C7H16INO2 | 273.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2260-50-6
Molecular Formula C7H16INO2
Molecular Weight (g/mol) 273.11
MDL Number MFCD00011815
InChI Key SMBBQHHYSLHDHF-UHFFFAOYSA-M
Synonym acetylcholine iodide, acetylcolina, 2-acetoxy-n,n,n-trimethylethanaminium iodide, acetylcolina italian, unii-7zcp12s7hq, choline, iodide, acetate ester, 2-acetoxyethyl trimethylammonium iodide, 7zcp12s7hq, ethanaminium, 2-acetyloxy-n,n,n-trimethyl-, iodide, 2-acetyloxy-n,n,n-trimethylethanamium iodide
PubChem CID 75271
SMILES [I-].CC(=O)OCC[N+](C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 162.0°C to 164.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3COOCH2CH2N(CH3)3I
Quantity 10 g
Solubility Information Solubility in water: negligible.
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;iodide
Formula Weight 273.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Acetylcholine iodide, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-862-3
RTECSNumber KH3300000
TSCA TSCA

RUO – Research Use Only