missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine Chloride 98.0+%, TCI America™

Catalog No. A008425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A008425G 25 g
A0084500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A008425G Supplier TCI America Supplier No. A008425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 60-31-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00011698
InChI Key JUGOREOARAHOCO-UHFFFAOYSA-M
Synonym acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
PubChem CID 6060
ChEBI CHEBI:2417
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
SMILES CC(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Melting Point 150°C
Color White
pH 4.0 to 5.7 (100g/L 25°C)
Quantity 25 g
Formula Weight 181.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Acetylcholine Chloride
EINECSNumber (9)-0249&(9)-1504
RTECSNumber FZ9800000
TSCA Yes