missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetylcholine Chloride Crystalline MP Biomedicals
Specifications
Specifications
| Quantity | 100 g |
| Formula Weight | 181.7 |
| Percent Purity | ∼99% |
| Physical Form | Crystalline |
| Chemical Name or Material | Acetylcholine Chloride |