missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetylcholine chloride, ∽99%, MP Biomedicals™
Acetylcholine chloride is a cholinergic neurotransmitter that can induce the opening of calcium channels, as well as act on nicotinic and muscarinic acetylcholine receptors
Manufacturer: Mp Biomedicals 0210001225
Specifications
Acetylcholine chloride | |
25g | |
∽99% | |
MFCD00011698 | |
JUGOREOARAHOCO-UHFFFAOYSA-M | |
2-acetyloxyethyl(trimethyl)azanium;chloride | |
6060 | |
181.7 |
White | |
60-31-1 | |
C7H16ClNO2 | |
2-acetyloxy-n,n,n-trimethylethanaminium chloride, acecholin, acecoline, acetylcholine chloride, ach chloride, arterocoline, azetylcholinchlorid, chloroacetylcholine, miochol, ovisot | |
CC(=O)OCC[N+](C)(C)C.[Cl-] | |
181.66 | |
CHEBI:2417 | |
Crystalline Powder |
Chemical Identifiers
60-31-1 | |
181.66 | |
JUGOREOARAHOCO-UHFFFAOYSA-M | |
6060 | |
2-acetyloxyethyl(trimethyl)azanium;chloride |
C7H16ClNO2 | |
MFCD00011698 | |
2-acetyloxy-n,n,n-trimethylethanaminium chloride, acecholin, acecoline, acetylcholine chloride, ach chloride, arterocoline, azetylcholinchlorid, chloroacetylcholine, miochol, ovisot | |
CHEBI:2417 | |
CC(=O)OCC[N+](C)(C)C.[Cl-] |
Safety and Handling
EINECSNumber : 200-468-8