missing translation for 'onlineSavingsMsg'
Learn More

Acetylcholine chloride, ∽99%, MP Biomedicals™

Catalog No. p-4403606
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10001225 25 g
ICN10001290 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10001225 Supplier MP Biomedicals Inc Supplier No. 0210001225
Only null left

Acetylcholine chloride is a cholinergic neurotransmitter that can induce the opening of calcium channels, as well as act on nicotinic and muscarinic acetylcholine receptors

Chemical Identifiers

CAS 60-31-1
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00011698
InChI Key JUGOREOARAHOCO-UHFFFAOYSA-M
Synonym acetylcholine chloride, miochol, acecoline, chloroacetylcholine, arterocoline, acecholin, ovisot, ach chloride, azetylcholinchlorid, 2-acetyloxy-n,n,n-trimethylethanaminium chloride
PubChem CID 6060
ChEBI CHEBI:2417
IUPAC Name 2-acetyloxyethyl(trimethyl)azanium;chloride
SMILES CC(=O)OCC[N+](C)(C)C.[Cl-]

Specifications

Color White
Assay Percent Range ∽99%
Quantity 25 g
Formula Weight 181.7
Physical Form Crystalline Powder
Chemical Name or Material Acetylcholine chloride
EINECSNumber 200-468-8